Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053991.1 | NP_446443.1 | 3 | 295 | ||
XM_006227827.2 | XP_006227889.1 | 4 | 740 | |||
XM_006227828.2 | XP_006227890.1 | 4 | 743 | |||
GenBank mRNA | AB089807.1 | - | 3 | 217 | ||
AB089808.1 | - | 2 | 452 | |||
AB089809.1 | - | 3 | 312 | |||
BC158798.1 | - | 3 | 295 | 7316540 | ||
CB583853.1 | - | 3 | 309 | |||
X02341.1 | - | 2 | 130 |