Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766363.1 | XP_008764585.1 | 16, 17 | 2109 | ||
XM_008766364.1 | XP_008764586.1 | 16, 17 | 2214 | |||
XM_008766365.1 | XP_008764587.1 | 16, 17 | 2091 |