Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237395.2 | XP_006237457.1 | 12 | 1955 | ||
XM_006237396.2 | XP_006237458.1 | 12 | 1955 | |||
XM_006237397.2 | XP_006237459.1 | 12 | 1954 | |||
XM_006237398.2 | XP_006237460.1 | 12 | 1954 | |||
XM_006237399.2 | XP_006237461.1 | 12 | 1933 | |||
XM_006237400.2 | XP_006237462.1 | 12 | 1915 | |||
XM_006237401.2 | XP_006237463.1 | 12 | 1954 | |||
XM_232354.8 | XP_232354.5 | 12 | 1933 |