Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057210.2 | NP_476558.2 | 10 | 1782 | ||
XM_006232952.2 | XP_006233014.1 | 11 | 2211 | |||
GenBank mRNA | BC092132.1 | - | 10 | 1782 | 7314271 | |
L05435.1 | - | 11 | 2128 |