Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201334.1 | NP_001188263.1 | 14, 15 | 1855 | ||
NM_003203.4 | NP_003194.3 | 14, 15 | 2087 | |||
XM_005264520.3 | XP_005264577.1 | 14, 15 | 2112 | |||
XM_011533074.2 | XP_011531376.1 | 13, 14 | 1863 | |||
GenBank mRNA | AB026911.1 | - | 14, 15 | 2025 | ||
AK097564.1 | - | 14, 15 | 1855 | |||
BC064559.1 | - | 14, 15 | 1984 | 5548897 | ||
EF158468.1 | - | 13, 14 | 1901 | |||
EF158469.1 | - | 10, 11 | 1301 |