Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080401.1 | NP_536326.1 | 3 | 783 | ||
XM_006229718.2 | XP_006229780.1 | 3 | 635 | |||
XM_006229719.2 | XP_006229781.1 | 4 | 1084 | |||
XM_006229720.2 | XP_006229782.1 | 4 | 2006 |