Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130411.2 | NP_569095.1 | 10, 11 | 1377 | ||
XM_006230204.2 | XP_006230266.1 | 10, 11 | 1461 | |||
GenBank mRNA | AF416730.1 | - | 9, 10 | 1273 | ||
AF495469.1 | - | 10, 11 | 1377 | |||
BC086971.1 | - | 10, 11 | 1353 | 7124429 | ||
FQ225277.1 | - | 7, 8 | 899 | |||
FQ225360.1 | - | 7, 8 | 899 | |||
FQ225522.1 | - | 6, 7 | 816 | |||
FQ226290.1 | - | 8, 9 | 846 | |||
FQ226379.1 | - | 10, 11 | 1368 | |||
FQ226941.1 | - | 10, 11 | 1397 | |||
FQ233306.1 | - | 10, 11 | 1371 | |||
FQ233390.1 | - | 10, 11 | 1373 | |||
FQ233414.1 | - | 10, 11 | 1375 | |||
FQ233980.1 | - | 10, 11 | 1291 | |||
FQ234750.1 | - | 10, 11 | 1379 |