Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173293.1 | NP_775415.1 | 2 | 1279 | ||
XM_008759669.1 | XP_008757891.1 | 3 | 2736 | |||
XM_008759670.1 | XP_008757892.1 | 3 | 1514 | |||
XM_008759671.1 | XP_008757893.1 | 3 | 943 | |||
GenBank mRNA | AF079864.1 | - | 2 | 1279 |