Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003211.4 | NP_003202.3 | 3 | 558 | ||
XM_005269125.1 | XP_005269182.1 | 3 | 418 | |||
XM_011538714.1 | XP_011537016.1 | 2 | 388 | |||
XM_011538715.1 | XP_011537017.1 | 2 | 388 | |||
GenBank mRNA | AK295387.1 | - | 1 | 45 | ||
AK299832.1 | - | 3 | 470 | |||
AK303631.1 | - | 2 | 385 | |||
AK309792.1 | - | 4 | 602 | |||
AK311673.1 | - | 2 | 388 | |||
AK313234.1 | - | 3 | 501 | |||
AL710869.1 | - | 4 | 710 | |||
BC019925.1 | - | 3 | 512 | |||
BC037557.1 | - | 3 | 511 | 5184772 | ||
DB162106.1 | - | 3 | 559 |