Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021961.5 | NP_068780.2 | 5 | 927 | ||
GenBank mRNA | AK310928.1 | - | 3 | 473 | ||
AL035973.1 | - | 3 | 212 | |||
AL833289.1 | - | 5 | 524 | |||
BC115398.1 | - | 4 | 420 | 40068609 |