Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133560.2 | NP_598244.2 | 13 | 1835 | ||
XM_006244954.2 | XP_006245016.1 | 13 | 1912 | |||
XM_008767072.1 | XP_008765294.1 | 13 | 1970 | |||
XM_008767073.1 | XP_008765295.1 | 13 | 2147 | |||
GenBank mRNA | AF474163.1 | - | 12 | 1465 | ||
AJ288898.2 | - | 13 | 1835 | |||
BC088393.1 | - | 13 | 1834 | 7124967 |