Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133560.2 | NP_598244.2 | 2 | 429 | ||
XM_006244954.2 | XP_006245016.1 | 2 | 506 | |||
XM_008767072.1 | XP_008765294.1 | 2 | 507 | |||
XM_008767073.1 | XP_008765295.1 | 2 | 684 | |||
GenBank mRNA | AF474163.1 | - | 1 | 59 | ||
AJ288898.2 | - | 2 | 429 | |||
BC088393.1 | - | 2 | 428 | 7124967 |