Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145398.2 | NP_001138870.1 | 2, 3 | 588 | ||
NM_003216.3 | NP_003207.1 | 2, 3 | 594 | |||
GenBank mRNA | AK091916.1 | - | 2, 3 | 466 | ||
AK295595.1 | - | 2, 3 | 520 | |||
AK314906.1 | - | 2, 3 | 510 | |||
BC039258.1 | - | 2, 3 | 563 | 5286096 | ||
BC042476.2 | - | 2, 3 | 564 | 4915095 | ||
CR456592.1 | - | 2, 3 | 491 | |||
CR541827.1 | - | 2, 3 | 478 | |||
DA243618.1 | - | 2, 3 | 533 | |||
U06935.1 | - | 2, 3 | 402 | |||
U44059.1 | - | 2, 3 | 514 |