Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134339.1 | NP_599166.1 | 1 | 196 | ||
GenBank mRNA | L13207.1 | - | 1 | 13 |