Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UST4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134379.1 | NP_599206.1 | 1 | 389 | ||
GenBank mRNA | AJ132859.1 | - | 1 | 389 | ||
BC100065.1 | - | 279 | 7373541 | |||
FQ210050.1 | - | 310 | ||||
FQ218143.1 | - | 449 |
Gene Symbol: Ust5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134380.1 | NP_599207.1 | 1 | 479 | ||
XM_008760213.1 | XP_008758435.1 | 1 | 1092 | |||
GenBank mRNA | AJ132858.1 | - | 1 | 479 | ||
BC081730.1 | - | 1 | 798 | |||
BC098905.1 | - | 1 | 455 | 7325342 |