Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138905.2 | NP_620260.2 | 3 | 775 | ||
GenBank mRNA | BC072544.1 | - | 3 | 775 | 7105872 | |
Y07783.1 | - | 3 | 767 |