Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191639.1 | NP_001178568.1 | 2754 | |||
GenBank mRNA | Y15054.1 | - | 1204 |