Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013036.1 | NP_001013054.1 | 6 | 560 | ||
XM_006251925.2 | XP_006251987.1 | 6 | 621 | |||
XM_006251926.2 | XP_006251988.1 | 6 | 621 | |||
XM_006251927.2 | XP_006251989.1 | 7 | 716 | |||
GenBank mRNA | BC089848.1 | - | 6 | 560 | 7374684 | |
FQ211507.1 | - | 7 | 656 |