Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003239.3 | NP_003230.1 | 1 | 1422 | ||
GenBank mRNA | AK313564.1 | - | 1 | 503 | ||
BC018503.1 | - | 1 | 393 | 4477175 | ||
BT007287.1 | - | 1 | 293 | |||
EF560714.1 | - | 1 | 406 | |||
J03241.1 | - | 1 | 555 | |||
X14149.1 | - | 1 | 546 |