Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144743.1 | NP_653344.1 | 1082 | |||
GenBank mRNA | AY034877.1 | - | 1082 |
Gene Symbol: Ces2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760594.1 | XP_008758816.1 | 5 | 737 |