Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145088.1 | NP_659556.1 | 5 | 2167 | ||
GenBank mRNA | AF308611.1 | - | 5 | 2167 |