Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022037.2 | NP_071320.2 | 7 | 753 | ||
NM_022173.2 | NP_071505.2 | 8 | 786 | |||
XM_005264527.1 | XP_005264584.1 | 7 | 763 | |||
XM_005264528.4 | XP_005264585.1 | 7 | 698 | |||
XM_005264531.1 | XP_005264588.1 | 9 | 833 | |||
XM_011533077.1 | XP_011531379.1 | 8 | 796 | |||
XM_011533078.1 | XP_011531380.1 | 7 | 4459 | |||
XM_011533081.2 | XP_011531383.1 | 8 | 1559 | |||
XM_011533082.2 | XP_011531384.1 | 6 | 5131 | |||
XM_017004789.1 | XP_016860278.1 | 8 | 615 | |||
XM_017004790.1 | XP_016860279.1 | 7 | 581 | |||
XM_017004791.1 | XP_016860280.1 | 6 | 665 | |||
XM_017004792.1 | XP_016860281.1 | 7 | 2970 | |||
XM_017004793.1 | XP_016860282.1 | 7 | 2795 | |||
XM_017004794.1 | XP_016860283.1 | 7 | 1527 | |||
XM_017004795.1 | XP_016860284.1 | 9 | 793 | |||
XM_017004796.1 | XP_016860285.1 | 7 | 4301 | |||
XM_017004797.1 | XP_016860286.1 | 8 | 2097 | |||
XM_017004798.1 | XP_016860287.1 | 8 | 828 | |||
XM_017004799.1 | XP_016860288.1 | 8 | 790 | |||
XM_017004800.1 | XP_016860289.1 | 7 | 757 | |||
GenBank mRNA | AB209211.1 | - | 8 | 797 | ||
BC015944.1 | - | 8 | 593 | 3909177 | ||
CN295707.1 | - | 7 | 669 | |||
DR157578.1 | - | 2 | 125 | |||
M77142.1 | - | 7 | 592 |