Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homez | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152849.2 | NP_690062.3 | 2 | 1068 | ||
XM_006251930.2 | XP_006251992.2 | 2 | 1918 | |||
XM_008770663.1 | XP_008768885.1 | 2 | 1915 | |||
XM_008770664.1 | XP_008768886.1 | 3 | 1403 | |||
GenBank mRNA | AY126016.1 | - | 1 | 986 | ||
BC084701.1 | - | 2 | 1068 | 7133916 | ||
BC085694.1 | - | 2 | 1053 | 7135570 |