Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153313.1 | NP_695225.1 | 4 | 695 | ||
GenBank mRNA | AB008422.1 | - | 4 | 695 | ||
BC078696.1 | - | 4 | 702 | 7126263 | ||
FQ219613.1 | - | 4 | 745 | |||
J02867.1 | - | 4 | 695 | |||
M16654.1 | - | 4 | 695 | |||
M22328.1 | - | 4 | 711 |