Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003256.3 | NP_003247.1 | 2 | 729 | ||
GenBank mRNA | AK313018.1 | - | 2 | 290 | ||
BC010553.1 | - | 2 | 729 | 4154090 | ||
BT019627.1 | - | 2 | 218 | |||
U76456.1 | - | 2 | 277 |