Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003266.3 | NP_003257.1 | 4 | 955 | ||
NM_138554.4 | NP_612564.1 | 3 | 835 | |||
NM_138557.2 | NP_612567.1 | 2 | 668 | |||
GenBank mRNA | AB445638.1 | - | 3 | 537 | ||
AK226155.1 | - | 4 | 710 | |||
AK290053.1 | - | 4 | 938 | |||
AK293068.1 | - | 3 | 666 | |||
AK303730.1 | - | 2 | 499 | |||
BC025294.1 | - | 2 | 454 | |||
BC117422.1 | - | 3 | 557 | 40125973 | ||
BC143841.1 | - | 4 | 677 | |||
U88880.1 | - | 4 | 701 | |||
U93091.1 | - | 3 | 640 |