Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173332.2 | NP_775454.2 | 7 | 1895 | ||
XM_006237141.2 | XP_006237203.1 | 8 | 2418 | |||
XM_006237142.2 | XP_006237204.1 | 7 | 3474 | |||
XM_006237143.2 | XP_006237205.1 | 7 | 2272 | |||
XM_006237145.2 | XP_006237207.1 | 8 | 2735 | |||
XM_006237146.1 | XP_006237208.1 | 5 | 1576 | |||
XM_008763224.1 | XP_008761446.1 | 8 | 2736 | |||
XM_008763225.1 | XP_008761447.1 | 9 | 2489 | |||
XM_008763226.1 | XP_008761448.1 | 7 | 2589 | |||
XM_008763227.1 | XP_008761449.1 | 5 | 1703 | |||
GenBank mRNA | BC162051.1 | - | 7 | 1895 | 7112072 |