Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191589.1 | NP_001178518.1 | 6 | 873 | ||
GenBank mRNA | FQ214717.1 | - | 6 | 892 | ||
FQ215051.1 | - | 6 | 855 | |||
FQ216207.1 | - | 6 | 855 | |||
FQ216222.1 | - | 6 | 855 | |||
FQ216295.1 | - | 6 | 867 | |||
FQ216388.1 | - | 6 | 893 | |||
FQ216645.1 | - | 6 | 854 | |||
FQ216791.1 | - | 6 | 893 | |||
FQ224678.1 | - | 6 | 902 |