Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105783.1 | NP_001099253.1 | 3 | 690 | ||
XM_006246448.2 | XP_006246510.1 | 3 | 700 |