Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191642.1 | NP_001178571.1 | 6 | 3011 | ||
XM_006246453.2 | XP_006246515.1 | 7 | 3453 | |||
XM_006246455.2 | XP_006246517.1 | 6 | 3054 | |||
XM_006246457.2 | XP_006246519.1 | 6 | 3114 |