Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021109.3 | NP_066932.1 | 1 | 430 | ||
GenBank mRNA | AK055976.1 | - | 2 | 1504 | ||
AK057827.1 | - | 3 | 427 | |||
AV715954.1 | - | 3 | 430 | |||
BC001631.1 | - | 1 | 411 | |||
BC016732.1 | - | 1 | 431 | |||
BC022857.1 | - | 1 | 431 | |||
BC061586.1 | - | 3 | 417 | |||
BC062436.1 | - | 1 | 431 | |||
BC064954.1 | - | 3 | 411 | |||
BC070248.1 | - | 3 | 428 | |||
BC070249.1 | - | 1 | 427 | |||
BC083509.1 | - | 1 | 427 | |||
BC092437.1 | - | 1 | 427 | |||
BC104464.1 | - | 1 | 381 | 40031330 | ||
BC104465.1 | - | 1 | 381 | 40031337 | ||
BC104666.1 | - | 3 | 431 | |||
BC107789.1 | - | 3 | 434 | |||
BC139925.1 | - | 1 | 416 | 40148985 | ||
BC151215.1 | - | 1 | 417 | 40148015 | ||
M11948.1 | - | 1 | 227 | |||
M17733.1 | - | 3 | 428 | |||
X02493.1 | - | 2 | 331 |