Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039378.1 | NP_001034467.1 | 4 | 283 | ||
GenBank mRNA | AY903234.1 | - | 4 | 283 | ||
AY903235.1 | - | 4 | 283 | |||
AY903236.1 | - | 4 | 283 | |||
AY903237.1 | - | 4 | 283 | |||
AY903238.1 | - | 4 | 283 | |||
BC166871.1 | - | 1 | 733 |