Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkiras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105839.1 | NP_001099309.1 | 2, 3 | 302 | ||
XM_006247274.2 | XP_006247336.1 | 2, 3 | 253 | |||
XM_008767974.1 | XP_008766196.1 | 2, 3 | 259 | |||
GenBank mRNA | BC168163.1 | - | 2, 3 | 196 | 8366401 |