Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005425.4 | NP_005416.1 | 1 | 366 | ||
GenBank mRNA | BC096135.3 | - | 1 | 344 | 40002547 | |
BC096136.3 | - | 1 | 344 | 40002550 | ||
BC096137.1 | - | 1 | 344 | 40002553 | ||
BX282422.1 | - | 1 | 366 | |||
X63758.1 | - | 1 | 325 | |||
X63759.1 | - | 1 | 593 |