Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248575.2 | XP_006248637.1 | 45 | 4235 | ||
XM_006248577.2 | XP_006248639.1 | 45 | 4227 | |||
XM_006248578.2 | XP_006248640.1 | 45 | 4259 | |||
XM_006248579.2 | XP_006248641.1 | 44 | 4229 | |||
XM_008768830.1 | XP_008767052.1 | 45 | 4434 |