Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003395.2 | NP_001003395.1 | 3 | 555 | ||
NM_001003396.2 | NP_001003396.1 | 3 | 453 | |||
NM_001003397.2 | NP_001003397.1 | 3 | 453 | |||
NM_001292026.2 | NP_001278955.1 | 3 | 555 | |||
NM_001300994.2 | NP_001287923.1 | 3 | 453 | |||
NM_001318903.1 | NP_001305832.1 | 3 | 453 | |||
NM_001318907.1 | NP_001305836.1 | 3 | 350 | |||
NM_003287.3 | NP_003278.1 | 3 | 453 | |||
XM_005267122.2 | XP_005267179.1 | 3 | 234 | |||
XM_006715557.3 | XP_006715620.1 | 3 | 699 | |||
XM_017011237.1 | XP_016866726.1 | 3 | 699 | |||
XM_017011238.1 | XP_016866727.1 | 3 | 699 | |||
XM_017011239.1 | XP_016866728.1 | 3 | 555 | |||
XM_017011240.1 | XP_016866729.1 | 3 | 233 | |||
GenBank mRNA | AF004427.1 | - | 3 | 270 | ||
AF208012.1 | - | 3 | 269 | |||
AK124817.1 | - | 3 | 555 | |||
AK291866.1 | - | 3 | 417 | |||
AK291872.1 | - | 3 | 287 | |||
BC002375.2 | - | 3 | 343 | 2822877 | ||
BE409865.1 | - | 3 | 481 | |||
BG770463.1 | - | 3 | 351 | |||
BM048527.1 | - | 3 | 473 | |||
BM546629.1 | - | 3 | 326 | |||
BP334548.1 | - | 3 | 369 | |||
BQ641312.1 | - | 3 | 332 | |||
CB114736.1 | - | 3 | 453 | |||
U44427.1 | - | 3 | 410 |