Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aimp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037348.1 | NP_001032425.1 | 3 | 407 | ||
XM_006248863.2 | XP_006248925.1 | 3 | 354 | |||
GenBank mRNA | BC107947.1 | - | 3 | 407 | 7386574 |