Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127246.1 | NP_001120718.1 | 5 | 2039 | ||
XM_006248998.2 | XP_006249060.1 | 5 | 2752 | |||
XM_006248999.2 | XP_006249061.1 | 5 | 2596 | |||
XM_006249000.2 | XP_006249062.1 | 5 | 2687 | |||
GenBank mRNA | BC166722.1 | - | 5 | 2039 | 7933458 |