Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286272.1 | NP_001273201.1 | 3 | 429 | ||
NM_001286273.1 | NP_001273202.1 | 2 | 355 | |||
NM_003295.3 | NP_003286.1 | 3 | 429 | |||
GenBank mRNA | AF072098.1 | - | 309 | |||
AK130847.1 | - | 3 | 328 | |||
AK296587.1 | - | 3 | 328 | |||
AK296952.1 | - | 2 | 855 | |||
AK312951.1 | - | 3 | 328 | |||
AY117678.1 | - | 3 | 329 | |||
AY334563.1 | - | 3 | 214 | |||
BC003352.2 | - | 3 | 312 | 2899964 | ||
BC012431.1 | - | 3 | 288 | 3880559 | ||
BC022436.1 | - | 2 | 283 | 4281364 | ||
BC040008.1 | - | 2 | 225 | |||
BC052333.1 | - | 3 | 318 | 6058799 | ||
BP215975.1 | - | 3 | 430 | |||
CR457036.1 | - | 3 | 214 | |||
L13806.1 | - | 1 | 49 | |||
X16064.1 | - | 3 | 308 | |||
X64899.1 | - | 3 | 300 |