Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rita1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044226.1 | NP_001037691.1 | 4 | 1423 | ||
XM_006249383.2 | XP_006249445.1 | 4 | 1526 | |||
XM_006249384.2 | XP_006249446.1 | 3 | 1401 | |||
GenBank mRNA | BC112384.1 | - | 4 | 1423 | 7932995 |