Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iscu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105936.1 | NP_001099406.1 | 1, 2 | 163 | ||
GenBank mRNA | BC167056.1 | - | 1, 2 | 142 | 7369214 |