Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010947.1 | NP_001010947.1 | 4 | 930 | ||
GenBank mRNA | BC089060.1 | - | 4 | 930 | 7320396 | |
FQ213644.1 | - | 4 | 922 |