Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134739.1 | NP_001128211.1 | 9 | 1717 | ||
NM_001134740.1 | NP_001128212.1 | 9 | 1717 | |||
XM_006250037.2 | XP_006250099.1 | 10 | 1981 | |||
XM_006250038.2 | XP_006250100.1 | 10 | 1799 | |||
XM_006250039.2 | XP_006250101.1 | 10 | 1750 | |||
XM_008769615.1 | XP_008767837.1 | 10 | 1768 | |||
XM_008769616.1 | XP_008767838.1 | 10 | 1935 | |||
GenBank mRNA | BC168218.1 | - | 9 | 1717 | 7319314 |