Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105976.1 | NP_001099446.1 | 4 | 484 | ||
GenBank mRNA | FQ217742.1 | - | 4 | 291 | ||
FQ229252.1 | - | 4 | 246 |