Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320350.1 | NP_001307279.1 | 18 | 2843 | ||
NM_001320351.1 | NP_001307280.1 | 18 | 2843 | |||
NM_003307.3 | NP_003298.1 | 18 | 2843 | |||
NR_038257.1 | 18 | 2846 | ||||
XM_005261171.3 | XP_005261228.1 | 19 | 3004 | |||
XM_011529736.2 | XP_011528038.1 | 18 | 3247 | |||
XM_017028456.1 | XP_016883945.1 | 19 | 3007 | |||
XM_017028457.1 | XP_016883946.1 | 18 | 3248 | |||
GenBank mRNA | AB001535.1 | - | 18 | 3188 | ||
AB166745.1 | - | 19 | 2956 | |||
AB166746.1 | - | 14 | 2195 | |||
AJ417076.1 | - | 19 | 2696 | |||
AJ878416.1 | - | 18 | 2806 | |||
BC112342.1 | - | 18 | 2756 | 40069893 |