Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282902.2 | NP_001269831.1 | 3 | 1623 | ||
NM_001282903.2 | NP_001269832.1 | 4 | 2112 | |||
NM_014112.4 | NP_054831.2 | 4 | 2126 | |||
XM_005251049.2 | XP_005251106.1 | 3 | 1941 | |||
XM_006716625.1 | XP_006716688.1 | 4 | 2063 | |||
XM_011517264.1 | XP_011515566.1 | 5 | 2196 | |||
XM_011517266.2 | XP_011515568.1 | 4 | 1931 | |||
XM_011517267.1 | XP_011515569.1 | 4 | 2049 | |||
XM_011517268.1 | XP_011515570.1 | 4 | 2038 | |||
XM_017013801.1 | XP_016869290.1 | 3 | 5725 | |||
GenBank mRNA | AF183810.1 | - | 4 | 2120 | ||
AF264784.1 | - | 3 | 1667 | |||
AK000948.1 | - | 1 | 110 | |||
AK304046.1 | - | 3 | 1618 | |||
AK309357.1 | - | 3 | 1837 | |||
BC125020.1 | - | 4 | 1956 | 40073132 |