Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105995.1 | NP_001099465.1 | 7 | 1599 | ||
XM_008770004.1 | XP_008768226.1 | 9 | 1934 | |||
XM_008770005.1 | XP_008768227.1 | 9 | 1838 | |||
XM_008770006.1 | XP_008768228.1 | 9 | 1776 | |||
XM_008770007.1 | XP_008768229.1 | 8 | 1603 |