Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191597.1 | NP_001178526.1 | 2 | 299 | ||
XM_006251031.2 | XP_006251093.1 | 2 | 303 |