Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fignl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011913.1 | NP_001011913.1 | 3 | 601 | ||
XM_006251476.2 | XP_006251538.1 | 3 | 624 | |||
XM_006251477.2 | XP_006251539.1 | 3 | 834 | |||
XM_008770261.1 | XP_008768483.1 | 3 | 648 | |||
GenBank mRNA | AY623031.1 | - | 3 | 601 | ||
AY623032.1 | - | 3 | 553 |