Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106027.1 | NP_001099497.1 | 8, 9 | 753 | ||
XM_006251752.2 | XP_006251814.1 | 8, 9 | 1458 | |||
XM_006251753.2 | XP_006251815.1 | 8, 9 | 778 | |||
XM_008770502.1 | XP_008768724.1 | 3, 4 | 1711 | |||
XM_008770503.1 | XP_008768725.1 | 6, 7 | 1164 | |||
XM_008770504.1 | XP_008768726.1 | 7, 8 | 882 | |||
XM_008770505.1 | XP_008768727.1 | 8, 9 | 2386 |